Learn More
Selleck Chemical LLC Tanespimycin (17-AAG) 100mg 75747-14-7 CP127374, NSC-330507, KOS 953
SDP
Catalog No. 508561
Encompass_Preferred
Tanespimycin (17-AAG, CP127374, NSC-330507, KOS 953) is a potent HSP90 inhibitor with IC50 of 5 nM in a cell-free assay, having a 100-fold higher binding affinity for HSP90 derived from tumour cells than HSP90 from normal cells. Tanespimycin (17-AAG) induces apoptosis, necrosis, autophagy and mitophagy. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-856-1 Supplier Selleck Chemical LLC Supplier No. S1141100MG
Tanespimycin (17-AAG, CP127374, NSC-330507, KOS 953) is a potent HSP90 inhibitor with IC50 of 5 nM in a cell-free assay, having a 100-fold higher binding affinity for HSP90 derived from tumour cells than HSP90 from normal cells. Tanespimycin (17-AAG) induces apoptosis, necrosis, autophagy and mitophagy. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.