Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 17-allylamino-17-demethoxygeldanamycin | 75747-14-7 | MFCD04973892 | 99.0% | 585.69 g/mol | C31H43N3O8 | 200 MG
SDP

Catalog No. 5000245823
Encompass_Preferred

Tanespimycin (17-AAG) is a benzoquinone-derived HSP90 inhibitor used as a research tool in cellular and biochemical studies of HSP90-dependent signaling and oncology models. It displays low-nanomolar potency against HSP90 and is provided in multiple pack formats for laboratory use.

  • Potent HSP90 inhibitor with an IC50 of 5 nM.
  • Preferential binding to tumor-derived HSP90 compared with normal HSP90.
  • Purity 99.01% as supplied.
  • Molecular formula C31H43N3O8; molecular weight 585.69 g/mol.
  • CAS number 75747-14-7.
  • Available as solid and solution formats, including a 200 mg pack.
  • Store protected from light; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-45823 Supplier Medchemexpress LLC Supplier No. HY10211200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tanespimycin (17-AAG) is a benzoquinone-derived HSP90 inhibitor used as a research tool in cellular and biochemical studies of HSP90-dependent signaling and oncology models. It displays low-nanomolar potency against HSP90 and is provided in multiple pack formats for laboratory use.

  • Potent HSP90 inhibitor with an IC50 of 5 nM.
  • Preferential binding to tumor-derived HSP90 compared with normal HSP90.
  • Purity 99.01% as supplied.
  • Molecular formula C31H43N3O8; molecular weight 585.69 g/mol.
  • CAS number 75747-14-7.
  • Available as solid and solution formats, including a 200 mg pack.
  • Store protected from light; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.