Learn More
Medchemexpress LLC 17-azido-3,6,9,12,15-pentaoxaheptadecan-1-ol | 86770-69-6 | MFCD22056309 | 98.0% | 307.34 g/mol | C12H25N3O6 | 1 G
SDP
Catalog No. 5000204263
Encompass_Preferred
Azido-PEG6-alcohol is a polyethylene glycol (PEG) linker bearing a terminal azide and a primary hydroxyl group, used as a click-chemistry reagent and linker in bioconjugation and PROTAC/ADC synthesis.
  • Water-soluble PEG6 linker with azide and hydroxyl functional groups.
  • Useful for copper-free and copper-catalyzed click reactions.
  • Commonly used as a spacer/linker in antibody-drug conjugates and PROTACs.
  • Molecular weight approximately 307.34 g/mol and formula C12H25N3O6.
  • Available in small-scale laboratory quantities (e.g., 1 G) and typically supplied with high purity.
Catalog No. 50-002-04263 Supplier Medchemexpress LLC Supplier No. HY1305371G
Azido-PEG6-alcohol is a polyethylene glycol (PEG) linker bearing a terminal azide and a primary hydroxyl group, used as a click-chemistry reagent and linker in bioconjugation and PROTAC/ADC synthesis.
  • Water-soluble PEG6 linker with azide and hydroxyl functional groups.
  • Useful for copper-free and copper-catalyzed click reactions.
  • Commonly used as a spacer/linker in antibody-drug conjugates and PROTACs.
  • Molecular weight approximately 307.34 g/mol and formula C12H25N3O6.
  • Available in small-scale laboratory quantities (e.g., 1 G) and typically supplied with high purity.