Learn More
Abcam 17 beta Estradiol ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1086671X96TESTS
Encompass_Preferred
The 17 beta Estradiol ELISA kit is a competitive ELISA designed to quantify 17-beta-estradiol (E2) with a sensitivity of 8.68 pg/mL. - High sensitivity – detects 17 beta estradiol (E2) as low as 8.68 pg/mL - Wide dynamic range – quantifies 20–2,000 pg/mL - Compatible with multiple sample types – works with serum, plasma (citrate, EDTA, heparin), and urine - Cited in over 55 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1000135