Learn More
Medchemexpress LLC 17-(Boc-amino)-3,6,9,12,15-pentaoxaheptadecan-1-oic acid | 890152-41-7 | 99.9% | 395.45 g/mol | C17H33NO9 | 100 MG
SDP
Catalog No. 5000283192
Encompass_Preferred
t-Boc-N-amido-PEG5-acetic acid is a Boc-protected PEG5-based linker that provides a flexible, hydrophilic spacer with a terminal carboxylic acid for use in PROTAC synthesis and bioconjugation. It is compatible with standard amide-forming chemistries and is supplied at research-grade purity with available batch COAs.
  • Provides a PEG5 hydrophilic spacer for reduced steric hindrance and improved solubility.
  • Contains a Boc-protected amine enabling selective deprotection and downstream functionalization.
  • Has a terminal carboxylic acid suitable for amide bond formation with ligands.
  • High purity and batch-specific COA available for quality assurance.
  • Offered in multiple package sizes for small-scale and preparative workflows.
Catalog No. 50-002-83192 Supplier Medchemexpress LLC Supplier No. HYW190962100MG
t-Boc-N-amido-PEG5-acetic acid is a Boc-protected PEG5-based linker that provides a flexible, hydrophilic spacer with a terminal carboxylic acid for use in PROTAC synthesis and bioconjugation. It is compatible with standard amide-forming chemistries and is supplied at research-grade purity with available batch COAs.
  • Provides a PEG5 hydrophilic spacer for reduced steric hindrance and improved solubility.
  • Contains a Boc-protected amine enabling selective deprotection and downstream functionalization.
  • Has a terminal carboxylic acid suitable for amide bond formation with ligands.
  • High purity and batch-specific COA available for quality assurance.
  • Offered in multiple package sizes for small-scale and preparative workflows.