Learn More
American Biotech Supply 17 CUBIC FOOT STANDARD AUTO DE
Catalog No. NC1884040
Encompass_Preferred
17 Cubic Foot Standard Auto Defrost Hydrocarbon Laboratory Freezer •Exterior dimensions: 28"W x 29 3/4"D x 68"H•Auto defrost•115V, 60Hz, 5 Amps, 1/4 HP•CSA listed•Shipping weight: 248 lbs.• DISCLAIMER: Auto defrost freezers incorporate an electric heater installed on the evaporator that assist in the removal of the frost and ice. Although the heater helps speed up the frost and ice removal, it can raise the temperatures within the freezer.
Catalog No. NC1884040 Supplier American Biotech Supply Supplier No. ABTHCAFS1720
17 Cubic Foot Standard Auto Defrost Hydrocarbon Laboratory Freezer •Exterior dimensions: 28"W x 29 3/4"D x 68"H•Auto defrost•115V, 60Hz, 5 Amps, 1/4 HP•CSA listed•Shipping weight: 248 lbs.• DISCLAIMER: Auto defrost freezers incorporate an electric heater installed on the evaporator that assist in the removal of the frost and ice. Although the heater helps speed up the frost and ice removal, it can raise the temperatures within the freezer.