Learn More
AdipoGen 17-DMAG
SDP
Supplier:  AdipoGen AGCN20540M005
Encompass_Preferred
Chemical. CAS 467214-20-6. Formula C32H48N4O8. MW 616.8. Less toxic, more potent synthetic derivative of geldanamycin Prod. No. BVT-0196 https//adipogen.com/bvt-0196-geldanamycin.html . Angiogenesis inhibitor. Heat shock protein 90 HSP90 inhibitor. Apoptosis inducer. Shows higher antitumor activity than 17-AAG Prod. No. BVT-0244 https//adipogen.com/bvt-0244-17-aag.html .
Catalog No. 50-256-2486