Learn More
Eurofins Discoverx 17 β-Estradiol, 10 mg. (92-1128)
Catalog No. 501999661
Encompass
β-Estradiol is an estrogen and the primary female sex hormone and steroid, acting as a regulator of the estrous and menstrual female reproductive cycles. Directs the development of the female organs in embryogenesis and during puberty. It is an estrogen receptor agonist (Ki=0.12 and 0.13 nM for ERα and ERβ, respectively). Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9661 Supplier Eurofins Discoverx Supplier No. 921128
β-Estradiol is an estrogen and the primary female sex hormone and steroid, acting as a regulator of the estrous and menstrual female reproductive cycles. Directs the development of the female organs in embryogenesis and during puberty. It is an estrogen receptor agonist (Ki=0.12 and 0.13 nM for ERα and ERβ, respectively). Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.