Learn More
Cayman Chemical 17-EstradIol-d2 500UG
SDP
Catalog No. 502892419
Encompass_Preferred
An internal standard for the quantification of 17β-estradiol by GC- or LC-MS
Catalog No. 50-289-2419 Supplier Cayman Chemical Supplier No. 9002846500 UG
An internal standard for the quantification of 17β-estradiol by GC- or LC-MS