Learn More
Cayman Chemical 17-EstradIol-d2 5mg
SDP
Catalog No. 502895724
Encompass_Preferred
An internal standard for the quantification of 17β-estradiol by GC- or LC-MS
Catalog No. 50-289-5724 Supplier Cayman Chemical Supplier No. 90028465 MG
An internal standard for the quantification of 17β-estradiol by GC- or LC-MS