Learn More
Cayman Chemical 17-EstradIol-d3 25mg
SDP
Catalog No. 502918501
Encompass_Preferred
An internal standard for the quantification of 17β-estradiol by GC- or LC-MS
Catalog No. 50-291-8501 Supplier Cayman Chemical Supplier No. 3897425 MG
An internal standard for the quantification of 17β-estradiol by GC- or LC-MS