Learn More
Cayman Chemical 17β-Estradiol-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 79037-37-9, 5 mg
SDP
Catalog No. 502918788
Encompass_Preferred
An internal standard for the quantification of 17β-estradiol by GC- or LC-MS
Catalog No. 50-291-8788 Supplier Cayman Chemical Supplier No. 389745 MG
An internal standard for the quantification of 17β-estradiol by GC- or LC-MS