Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 17β-Estradiol sulfate sodium | 4999-79-5 | 99.0% | C18H23NaO5S | 25 MG
SDP

Catalog No. 5000352915
Encompass_Preferred

17β-Estradiol sulfate sodium, also known as β-Estradiol 3-sulfate sodium salt, is a neuroactive steroid used for research applications. This compound is supplied as a white to light yellow solid with a high purity of 99.0%.

  • Neuroactive steroid
  • High purity of 99.0%
  • Supplied as a white to light yellow solid
  • Suitable for research applications
  • Store product at 4°C, sealed, and away from moisture
  • Store stock solutions at -80°C for up to 6 months, or -20°C for up to 1 month
  • Highly soluble in DMSO

Catalog No. 50-003-52915 Supplier Medchemexpress LLC Supplier No. 25MGHY141672
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

17β-Estradiol sulfate sodium, also known as β-Estradiol 3-sulfate sodium salt, is a neuroactive steroid used for research applications. This compound is supplied as a white to light yellow solid with a high purity of 99.0%.

  • Neuroactive steroid
  • High purity of 99.0%
  • Supplied as a white to light yellow solid
  • Suitable for research applications
  • Store product at 4°C, sealed, and away from moisture
  • Store stock solutions at -80°C for up to 6 months, or -20°C for up to 1 month
  • Highly soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.