Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 17β-estradiol sulfate sodium | 4999-79-5 | MFCD00056553 | 99.0% | 374.43 g·mol⁻¹ | C18H23NaO5S | 5 MG
SDP

Catalog No. 5000221746
Encompass_Preferred

17β-estradiol sulfate sodium is the sodium salt of the sulfate metabolite of 17β-estradiol supplied as a solid research reagent. It is used for studies of estrogen metabolism, receptor binding, and neuroactive steroid activity. For research use only; not for human or veterinary use.

  • Sodium salt of 17β-estradiol sulfate.
  • High purity (99.0%).
  • Molecular weight 374.43 g·mol⁻¹.
  • Chemical formula C18H23NaO5S.
  • Provided as a white to light yellow solid.
  • SDS and COA available for quality and safety documentation.

Catalog No. 50-002-21746 Supplier Medchemexpress LLC Supplier No. HY1416725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

17β-estradiol sulfate sodium is the sodium salt of the sulfate metabolite of 17β-estradiol supplied as a solid research reagent. It is used for studies of estrogen metabolism, receptor binding, and neuroactive steroid activity. For research use only; not for human or veterinary use.

  • Sodium salt of 17β-estradiol sulfate.
  • High purity (99.0%).
  • Molecular weight 374.43 g·mol⁻¹.
  • Chemical formula C18H23NaO5S.
  • Provided as a white to light yellow solid.
  • SDS and COA available for quality and safety documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.