Learn More
Medchemexpress LLC 17-hydroxy-11-dehydrocorticosterone | 53-06-5 | 99.3% | 360.44 | 5 G
SDP
Catalog No. 5000386261
Encompass_Preferred
Cortisone (17-Hydroxy-11-dehydrocorticosterone) is an oxidized metabolite of Cortisol, a Glucocorticoid. It functions as an immunosuppressant and anti-inflammatory agent. At high concentrations, Cortisone can partially interfere with the binding of Glucocorticoid to the Glucocorticoid-receptor. This product is intended for research use only.
  • Oxidized metabolite of Cortisol
  • Functions as an immunosuppressant
  • Acts as an anti-inflammatory agent
  • Partially interferes with Glucocorticoid-receptor binding
  • For research use only
Catalog No. 50-003-86261 Supplier Medchemexpress LLC Supplier No. HY174615G
Cortisone (17-Hydroxy-11-dehydrocorticosterone) is an oxidized metabolite of Cortisol, a Glucocorticoid. It functions as an immunosuppressant and anti-inflammatory agent. At high concentrations, Cortisone can partially interfere with the binding of Glucocorticoid to the Glucocorticoid-receptor. This product is intended for research use only.
  • Oxidized metabolite of Cortisol
  • Functions as an immunosuppressant
  • Acts as an anti-inflammatory agent
  • Partially interferes with Glucocorticoid-receptor binding
  • For research use only