Learn More
Cayman Chemical 17β-hydroxy Exemestane-d3, Purity: ≥99% deuterated forms (d1-d3), 1 mg
SDP
Catalog No. 502935598
Encompass_Preferred
An internal standard for the quantification of 17β-hydroxy exemestane and as a surrogate internal standard for related steroids such as exemestane by GC- or LC-MS
Catalog No. 50-293-5598 Supplier Cayman Chemical Supplier No. 257561 MG
An internal standard for the quantification of 17β-hydroxy exemestane and as a surrogate internal standard for related steroids such as exemestane by GC- or LC-MS