Learn More
Cayman Chemical 17-hydroxy Exemestn-d3 500ug
SDP
Catalog No. 502935864
Encompass_Preferred
An internal standard for the quantification of 17β-hydroxy exemestane and as a surrogate internal standard for related steroids such as exemestane by GC- or LC-MS
Catalog No. 50-293-5864 Supplier Cayman Chemical Supplier No. 25756500 UG
An internal standard for the quantification of 17β-hydroxy exemestane and as a surrogate internal standard for related steroids such as exemestane by GC- or LC-MS