Learn More
Cayman Chemical 17β-hydroxy Wortmannin, Purity: ≥98%, CAS: 58053-83-1, 10 mg
SDP
Catalog No. 502955088
Encompass_Preferred
An analog of wortmannin; irreversibly binds PI3K; inhibits recombinant PI3K and mTOR (IC50 = 2.7 and 193 nM, respectively) and prevents the growth of LNCap cells (IC50 = 1.46 μM)
Catalog No. 50-295-5088 Supplier Cayman Chemical Supplier No. 1381210 MG
An analog of wortmannin; irreversibly binds PI3K; inhibits recombinant PI3K and mTOR (IC50 = 2.7 and 193 nM, respectively) and prevents the growth of LNCap cells (IC50 = 1.46 μM)