Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 17-hydroxyheptadecanoic acid | 13099-34-8 | MFCD00506157 | 99.7% | 286.45 g/mol | C17H34O3 | 250MG
SDP

Catalog No. 5000239215
Encompass_Preferred

17-hydroxyheptadecanoic acid is a 17-carbon ω-hydroxy fatty acid supplied as a white to off-white solid intended for research use in lipid metabolism studies and analytical workflows. The material is accompanied by a batch certificate of analysis reporting 99.70% purity (HPLC), and includes molecular formula C17H34O3 and molecular weight 286.45 g/mol. Recommended storage: room temperature for the solid (up to 3 years); in solvent store at -80°C (2 years) or -20°C (1 year).

  • High purity suitable for analytical and research applications.
  • Batch certificate of analysis available for traceable quality.
  • Solid form is convenient for weighing and storage.
  • Storage conditions specified for solid and solvent forms.
  • Molecular formula and molecular weight provided for method development.
  • CAS number 13099-34-8 enables precise identification.

Catalog No. 50-002-39215 Supplier Medchemexpress LLC Supplier No. HY124282250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

17-hydroxyheptadecanoic acid is a 17-carbon ω-hydroxy fatty acid supplied as a white to off-white solid intended for research use in lipid metabolism studies and analytical workflows. The material is accompanied by a batch certificate of analysis reporting 99.70% purity (HPLC), and includes molecular formula C17H34O3 and molecular weight 286.45 g/mol. Recommended storage: room temperature for the solid (up to 3 years); in solvent store at -80°C (2 years) or -20°C (1 year).

  • High purity suitable for analytical and research applications.
  • Batch certificate of analysis available for traceable quality.
  • Solid form is convenient for weighing and storage.
  • Storage conditions specified for solid and solvent forms.
  • Molecular formula and molecular weight provided for method development.
  • CAS number 13099-34-8 enables precise identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.