Learn More
Apexbio Technology LLC 17-Hydroxyprogesterone 68-96-2 50mg
SDP
Catalog No. 502240320
Encompass_Preferred
17-Hydroxyprogesterone (68-96-2) is a small-molecule agonist targeting the progesterone receptor (PR) with partial agonist activity at glucocorticoid receptors (GR) and antagonist effects at mineralocorticoid receptors (MR) It modulates steroid hormone signaling pathways thereby influencing vascular tone and inflammation 17-Hydroxyprogesterone exerts its biological activity primarily through activation and modulation of steroid hormone receptors In animal models administration of 17-Hydroxyprogesterone demonstrates the ability to modulate vascular inflammation and uterine vascular tone Based on these pharmacological properties 17-Hydroxyprogesterone holds research potential in the screening of congenital adrenal hyperplasia and related metabolic syndromes as well as associations with respiratory distress syndrome and premature birth outcomes
This item is not returnable. View return policy
Catalog No. 50-224-0320 Supplier Apexbio Technology LLC Supplier No. B612650
17-Hydroxyprogesterone (68-96-2) is a small-molecule agonist targeting the progesterone receptor (PR) with partial agonist activity at glucocorticoid receptors (GR) and antagonist effects at mineralocorticoid receptors (MR) It modulates steroid hormone signaling pathways thereby influencing vascular tone and inflammation 17-Hydroxyprogesterone exerts its biological activity primarily through activation and modulation of steroid hormone receptors In animal models administration of 17-Hydroxyprogesterone demonstrates the ability to modulate vascular inflammation and uterine vascular tone Based on these pharmacological properties 17-Hydroxyprogesterone holds research potential in the screening of congenital adrenal hyperplasia and related metabolic syndromes as well as associations with respiratory distress syndrome and premature birth outcomes