Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 17-Hydroxyprogesterone Competitive ELISA Kit

Catalog No. EIA17OHP
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIA17OHP 96 Tests
EIA17OHPX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EIA17OHP Supplier Invitrogen™ Supplier No. EIA17OHP
Only null left
Add to Cart
Add to Cart

ELISA

The 17-Hydroxyprogesterone Competitive ELISA quantitates 17-Hydroxyprogesterone in serum,plasma,urine,dried fecal extracts or cell culture medium.

Principle of the method

The 17-Hydroxyprogesterone Competitive ELISA research-use-only kit uses a specifically generated antibody to measure 17-Hydroxyprogesterone (17HO-P) and its metabolites in reconstituted buffer samples,tissue culture media samples,urine and fecal samples,or in extracted serum and plasma. This kit is not recommended for serum or plasma samples without extraction. A 17HO-P standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into the provided plate coated with a capture antibody. A 17HO-P-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of a polyclonal antibody to 17HO-P to each well. After a 1-hour incubation the plate is washed and substrate is added. The substrate reacts with the bound 17HO-P-peroxidase conjugate. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.

Rigorous validation

Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.

17-Hydroxyprogesterone (4-pregen-17-ol-3, 20-dione, 17HO-P, OHPG) is a steroid hormone from the androgen group that is found in mammals, reptiles, birds, and other vertebrates. 17HO-P is identical across all species and so this kit should measure 17HO-P from all sources. In humans, it is primarily synthesized in the adrenal glands, the gonads, testes, and placenta.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 24.69 to 6000 pg/mL
Assay Sensitivity 20.3 pg/mL
Description 17-Hydroxyprogesterone Competitive ELISA Kit
Conjugate HRP
Product Type ELISA
Protein Family Steroid hormone
Sample Type Fecal Extract, Plasma, Serum, Supernatant, Urine
For Use With (Application) ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV 8%
Intraassay CV 6.6
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Reproductive Biology
Sample Volume Fecal Extract 50 μL, Plasma 50 μL, Serum 50 μL, Supernatant 50 μL, Urine 25 μL
Storage Requirements -20°C
Target 17-Hydroxyprogesterone
Test Time 1 hrs 20 mins
Total Assay Time 1 hrs 30 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.