Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedical™ 17α-Hydroxyprogesterone Double Antibody RIA Kit

Catalog No. MP07171102 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP07171102 500 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP07171102 Supplier MP Biomedical™ Supplier No. 07171102
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quantitative Liquid Phase Radioimmunoassay

The 17αHP Double Antibody RIA Kit is intended for the measurement of 17α-hydroxyprogesterone in unextracted serum or plasma of adults and children. The steroid 17α-hydroxyprogesterone (17αHP) is produced by both the adrenal cortex and gonads. Even though 17αHP has relatively little progestational activity, it is of intense clinical interest because it is the immediate precursor to 11-desoxycortisol (Cpd-S). Because Cpd-S is produced by 21-hydroxylation of 17αHP, measurement of 17αHP is a useful indirect indicator of 21-hydroxylase activity. In congenital 21-hydroxylase deficiency, the most common variety of congenital adrenal hyperplasia (CAH), 17αHP is secreted in abundant excess. It is moderately elevated in the 11-b-hydroxylase deficiency as well. Measurement of 17αHP is therefore valuable in the initial diagnosis of CAH.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Kit and Assay
Sample Type Plasma, Serum
Target Species Human
For Use With (Application) Endocrinology
Quantity 500 Tubes
Regulatory Status FDA Cleared (K840886)
Sample Volume 25 μL
Storage Requirements 2°C to 8°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.