Learn More
AdipoGen 17-Hydroxyventuricidin A (1 mg)
SDP
Supplier:  AdipoGen AGCN20119M001
Encompass_Preferred
Chemical. CAS: 113204-43-6. Formula: C41H67NO12. MW: 766. Isolated from Streptomyces sp. Antifungal. Antibacterial against Gram-positive bacteria.
Catalog No. 50-205-3485