Learn More
Cayman Chemical 17-methyl Stearic Acid, Purity: ≥95%, CAS: 2724-59-6, 10 mg
SDP
Catalog No. 502934715
Encompass_Preferred
A methyl branched-chain fatty acid; has been found in murine meibomian glands, C. cornucopioides mushrooms, and Phyllobacterium species; has been found in the aerial parts of C. ladanifer and its concentration exhibits seasonal variation
Catalog No. 50-293-4715 Supplier Cayman Chemical Supplier No. 2676610 MG
A methyl branched-chain fatty acid; has been found in murine meibomian glands, C. cornucopioides mushrooms, and Phyllobacterium species; has been found in the aerial parts of C. ladanifer and its concentration exhibits seasonal variation