Learn More
Medchemexpress LLC 17-octadecynoic acid | 34450-18-5 | MFCD00077382 | 98.0% | 280.45 g/mol | C18H32O2 | 10 MG
SDP
Catalog No. 5000211649
Encompass_Preferred
17-Octadecynoic acid is a fatty acid alkyne that functions as a CYP450 ω-hydroxylase inhibitor and a click-chemistry reagent containing an alkyne group. It is used in biochemical assays and in vivo studies to probe CYP450-mediated metabolism and related physiological effects.
  • Inhibits CYP450 ω-hydroxylase activity.
  • Potent inhibitor of 20-HETE and related metabolite formation.
  • Contains alkyne group suitable for copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Validated for in vitro and in vivo experimental applications.
  • Provided with high purity for biochemical research.
Catalog No. 50-002-11649 Supplier Medchemexpress LLC Supplier No. HY10101610MG
17-Octadecynoic acid is a fatty acid alkyne that functions as a CYP450 ω-hydroxylase inhibitor and a click-chemistry reagent containing an alkyne group. It is used in biochemical assays and in vivo studies to probe CYP450-mediated metabolism and related physiological effects.
  • Inhibits CYP450 ω-hydroxylase activity.
  • Potent inhibitor of 20-HETE and related metabolite formation.
  • Contains alkyne group suitable for copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Validated for in vitro and in vivo experimental applications.
  • Provided with high purity for biochemical research.