Learn More
Cayman Chemical 17-Octadecynoic Acid, 34450-18-5, 10 mg
SDP
Supplier:  Cayman Chemical 9027010
Encompass_Preferred
A suicide inhibitor of LTB4 20-hydroxylase and renal CYP450 ω-hydroxylase
Catalog No. NC0476307