Learn More
Alpco Ltd 17-alpha Hydroxy Progesterone ELISA
SDP
Supplier:  Alpco Ltd 2017OHUE01
Encompass_Preferred
The 17-alpha Hydroxy Progesterone ELISA is for the quantitative determination of 17-alpha Hydroxy Progesterone in Serum.
FDA Registered. For In Vitro Diagnostic Use in the United States.
Catalog No. 50-751-3500