Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 17-Phenyl trinor PGF 1mg | 130209-76-6 | 1 MG
SDP

Catalog No. 5000227317
Encompass_Preferred

17-phenyl trinor PGF2α isopropyl ester is a prostaglandin F-series analog and FP receptor agonist supplied for research use as a chemical intermediate or pharmacology reference. It is provided in small mass packs suitable for assay development, synthesis, and analytical standards.

  • Research reagent and FP receptor agonist for pharmacology studies.
  • Suitable as a reference standard or synthetic intermediate.
  • CAS number 130209-76-6 for unambiguous identification.
  • Molecular formula C26H38O5; molecular weight 430.6 g/mol.
  • Available in small mass pack sizes for assay and synthesis.

Catalog No. 50-002-27317 Supplier Medchemexpress LLC Supplier No. HY116161A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

17-phenyl trinor PGF2α isopropyl ester is a prostaglandin F-series analog and FP receptor agonist supplied for research use as a chemical intermediate or pharmacology reference. It is provided in small mass packs suitable for assay development, synthesis, and analytical standards.

  • Research reagent and FP receptor agonist for pharmacology studies.
  • Suitable as a reference standard or synthetic intermediate.
  • CAS number 130209-76-6 for unambiguous identification.
  • Molecular formula C26H38O5; molecular weight 430.6 g/mol.
  • Available in small mass pack sizes for assay and synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.