Learn More
Medchemexpress LLC 17-Phenyl trinor PGF 25mg | 130209-76-6 | 430.58 g/mol | C26H38O5 | 25 MG
SDP
Catalog No. 5000229287
Encompass_Preferred
17-Phenyl trinor PGF2α isopropyl ester (CAS 130209-76-6) is a prostaglandin analogue and FP receptor agonist supplied for laboratory research. It is the isopropyl ester derivative of 17-phenyl trinor PGF2α, provided as a white to off-white solid with high reported purity for use in receptor pharmacology, biochemical assays, and method development.
  • High purity (~99.8%) suitable for research applications.
  • White to off-white solid for straightforward handling and storage.
  • Available in 25 mg packs for small-scale experiments.
  • Functions as an FP receptor agonist for pharmacological studies.
  • Molecular weight 430.58 g/mol; formula C26H38O5 for reference.
Catalog No. 50-002-29287 Supplier Medchemexpress LLC Supplier No. HY116161A25MG
17-Phenyl trinor PGF2α isopropyl ester (CAS 130209-76-6) is a prostaglandin analogue and FP receptor agonist supplied for laboratory research. It is the isopropyl ester derivative of 17-phenyl trinor PGF2α, provided as a white to off-white solid with high reported purity for use in receptor pharmacology, biochemical assays, and method development.
  • High purity (~99.8%) suitable for research applications.
  • White to off-white solid for straightforward handling and storage.
  • Available in 25 mg packs for small-scale experiments.
  • Functions as an FP receptor agonist for pharmacological studies.
  • Molecular weight 430.58 g/mol; formula C26H38O5 for reference.