Learn More
Medchemexpress LLC 17-Phenyl trinor PGF 5mg | 130209-76-6 | 430.6 g/mol | C26H38O5 | 5 MG
SDP
Catalog No. 5000229288
Encompass_Preferred
Isopropyl ester derivative of 17-phenyl trinor prostaglandin F2α used as a research reagent in prostaglandin pharmacology. The compound acts as a potent FP receptor agonist and is supplied at high purity for in vitro and pharmacological studies.
  • Isopropyl ester of 17-phenyl trinor PGF2α.
  • Potent FP receptor agonist for prostaglandin research.
  • High purity (≥98%).
  • Molecular weight 430.6 g/mol; formula C26H38O5.
  • Available in small research pack sizes such as 5 mg.
Catalog No. 50-002-29288 Supplier Medchemexpress LLC Supplier No. HY116161A5MG
Isopropyl ester derivative of 17-phenyl trinor prostaglandin F2α used as a research reagent in prostaglandin pharmacology. The compound acts as a potent FP receptor agonist and is supplied at high purity for in vitro and pharmacological studies.
  • Isopropyl ester of 17-phenyl trinor PGF2α.
  • Potent FP receptor agonist for prostaglandin research.
  • High purity (≥98%).
  • Molecular weight 430.6 g/mol; formula C26H38O5.
  • Available in small research pack sizes such as 5 mg.