Learn More
Cayman Chemical 17 R-ProtectIn D1 10ug
SDP
Catalog No. 502948847
Encompass_Preferred
An aspirin-triggered epimer of protectin D1; decreases leukotriene B4-induced transendothelial migration of human PMNs at 0.1-10 nM; reduces the recruitment of neutrophils in a mouse model of TNF-α-induced peritonitis at 0.01-10 ng
Catalog No. 50-294-8847 Supplier Cayman Chemical Supplier No. 3419810 UG
An aspirin-triggered epimer of protectin D1; decreases leukotriene B4-induced transendothelial migration of human PMNs at 0.1-10 nM; reduces the recruitment of neutrophils in a mouse model of TNF-α-induced peritonitis at 0.01-10 ng