Learn More
Cayman Chemical 17 R-ResolvIn D1 10ug
SDP
Catalog No. 502953036
Encompass_Preferred
An aspirin-triggered epimer of RvD1; reduces human leukocyte transendothelial migration (EC50 = ~30 nM), and limits leukocyte infiltration in a mouse model of peritonitis
Catalog No. 50-295-3036 Supplier Cayman Chemical Supplier No. 1306010 UG
An aspirin-triggered epimer of RvD1; reduces human leukocyte transendothelial migration (EC50 = ~30 nM), and limits leukocyte infiltration in a mouse model of peritonitis