Learn More
Cayman Chemical 17 R-ResolvIn D1 25ug
SDP
Catalog No. 502952713
Encompass_Preferred
An aspirin-triggered epimer of RvD1; reduces human leukocyte transendothelial migration (EC50 = ~30 nM), and limits leukocyte infiltration in a mouse model of peritonitis
Catalog No. 50-295-2713 Supplier Cayman Chemical Supplier No. 1306025 UG
An aspirin-triggered epimer of RvD1; reduces human leukocyte transendothelial migration (EC50 = ~30 nM), and limits leukocyte infiltration in a mouse model of peritonitis