Learn More
Cayman Chemical 17 R-ResolvIn D1-d5 100ug
SDP
Catalog No. 502951355
Encompass_Preferred
An inhibitor of Ras-mediated signaling that functions by dislodging Ras from the cell membrane thereby rendering it susceptible to proteolytic degradation inhibits the growth of human Ha-ras-transformed Rat1 fibroblasts with an IC50 value of 7 5 UM
Catalog No. 50-295-1355 Supplier Cayman Chemical Supplier No. 11183100 UG
An inhibitor of Ras-mediated signaling that functions by dislodging Ras from the cell membrane thereby rendering it susceptible to proteolytic degradation inhibits the growth of human Ha-ras-transformed Rat1 fibroblasts with an IC50 value of 7 5 UM