Learn More
Cayman Chemical 17 R-ResolvIn D1-d5 25ug
SDP
Catalog No. 502951071
Encompass_Preferred
An internal standard for the quantification of RvD1 by GC- or LC-MS
Catalog No. 50-295-1071 Supplier Cayman Chemical Supplier No. 1118325 UG
An internal standard for the quantification of RvD1 by GC- or LC-MS