Learn More
Cayman Chemical 17 RHDHA Fatty Acids and Deriv
SDP
Supplier:  Cayman Chemical 1000509950 UG
Encompass_Preferred
An aspirin-induced DHA metabolite; a precursor to resolvins; inhibits TNF-α-induced IL-1β expression in human glioma cells
Catalog No. 50-289-0885