Learn More
Cayman Chemical 17 RHDHA Fatty Acids and Deriv
SDP
Catalog No. 502891246
Encompass_Preferred
An aspirin-induced DHA metabolite; a precursor to resolvins; inhibits TNF-α-induced IL-1β expression in human glioma cells
Catalog No. 50-289-1246 Supplier Cayman Chemical Supplier No. 10005099100 UG
An aspirin-induced DHA metabolite; a precursor to resolvins; inhibits TNF-α-induced IL-1β expression in human glioma cells