Learn More
Cayman Chemical 17 RHDHA Fatty Acids and Deriv
Supplier: Cayman Chemical 10005099250 UG
An aspirin-induced DHA metabolite; a precursor to resolvins; inhibits TNF-α-induced IL-1β expression in human glioma cells