Learn More
Cayman Chemical 17 RHETE Fatty Acids and Deriv
SDP
Supplier:  Cayman Chemical 1001063750 UG
Encompass_Preferred
An isomer of a CYP450 metabolite of arachidonic acid that has no effect on proximal tubule ATPase activity.
Catalog No. 50-288-9622