Learn More
Cayman Chemical 17 RHETE Fatty Acids and Deriv
SDP
Catalog No. 502888586
Encompass_Preferred
An isomer of a CYP450 metabolite of arachidonic acid that has no effect on proximal tubule ATPase activity.
Catalog No. 50-288-8586 Supplier Cayman Chemical Supplier No. 1001063725 UG
An isomer of a CYP450 metabolite of arachidonic acid that has no effect on proximal tubule ATPase activity.