Learn More
Cayman Chemical 17 RRSolvIn D1 methyl Ste 10UG
SDP
Catalog No. 502893770
Encompass_Preferred
A methyl ester version of 17(R)-RvD1 which may act as a lipophilic prodrug form that will alter its distribution and pharmacokinetic properties
Catalog No. 50-289-3770 Supplier Cayman Chemical Supplier No. 900107210 UG
A methyl ester version of 17(R)-RvD1 which may act as a lipophilic prodrug form that will alter its distribution and pharmacokinetic properties