Learn More
Cayman Chemical 17 SHDHA Fatty Acids and Deriv
SDP
Supplier:  Cayman Chemical 10009799250 UG
Encompass_Preferred
A hydroxy fatty acid formed from DHA by 15-LO and is a precursor to 17(S)-resolvins; inhibits platelet 12-LO (IC50 = 0.4 μM); inhibits TNF-α-induced expression of IL1B in a human glial cell line (IC50 = ~0.5 nM); inhibits NLRP3 inflammasome formation induced by homocysteine in podocytes at 100 nM
Catalog No. 50-288-8439