Learn More
Cayman Chemical 17 SHETE Fatty Acids and Deriv
SDP
Catalog No. 502890667
Encompass_Preferred
A potent inhibitor of proximal tubule ATPase activity; is a CYP450 metabolite of arachidonic acid; inhibits proximal tubule ATPase activity by 70% at 2 µM
Catalog No. 50-289-0667 Supplier Cayman Chemical Supplier No. 10011305100 UG
A potent inhibitor of proximal tubule ATPase activity; is a CYP450 metabolite of arachidonic acid; inhibits proximal tubule ATPase activity by 70% at 2 µM