Learn More
Cayman Chemical 17 SHETE Fatty Acids and Deriv
Supplier: Cayman Chemical 1001130525 UG
A potent inhibitor of proximal tubule ATPase activity; is a CYP450 metabolite of arachidonic acid; inhibits proximal tubule ATPase activity by 70% at 2 µM