Learn More
Ace Glass, Inc. 170 X 130MM TEF SUPP RING 170 X 130MM TEF SUPP RING
SDP
Catalog No. 50980634
Encompass
170 x 130mm PTFE support ring with internal bevel. Support ring is useful for supporting columns, flasks, dishes, etc.
Catalog No. 50-980-634 Supplier Ace Glass, Inc. Supplier No. 1175208
170 x 130mm PTFE support ring with internal bevel. Support ring is useful for supporting columns, flasks, dishes, etc.