Learn More
CELLTREAT Scientific Products 1700cm2 Roller Bottle, Vent Cap, Sterile
SDP
Catalog No. NC1772261
Encompass_Preferred
CELLTREAT Scientific Products 1700cm2 Roller Bottles have uniform hydrophilic surface ideal for consisten cell attachment. Large knurled cap for easy grip. Easy-to-read black printed graduations
Catalog No. NC1772261 Supplier CELLTREAT Scientific Products Supplier No. 229375
CELLTREAT Scientific Products 1700cm2 Roller Bottles have uniform hydrophilic surface ideal for consisten cell attachment. Large knurled cap for easy grip. Easy-to-read black printed graduations