Learn More
Sino Biological Recombinant Human TL1A/TNFSF15 Protein (mFc Tag)
Supplier:  Sino Biological 17049H04H100
Encompass_Preferred
A DNA sequence encoding the human TNFSF15 (NP_005109.2) (Leu72-Leu251) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-161-9991