Learn More
Sino Biological Recombinant Human TL1A/TNFSF15 Protein (rFc Tag)
Supplier:  Sino Biological 17049H15H100
Encompass_Preferred
A DNA sequence encoding the human TNFSF15 (NP_005109.2) (Leu72-Leu251) was expressed with the Fc region of rabbit IgG at the N-terminus..
Catalog No. 50-161-9993