Learn More
Sino Biological Recombinant Human Factor B Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 17064H08H100
Encompass_Preferred
A DNA sequence encoding the human CFB (AAH04143.1) (Met1-Leu764) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9997