Learn More
Ace Glass, Inc. 170x90mm crystallization dish, cs/8, sp/2, 3140-170
SDP
Catalog No. 509558483
Encompass
170X90mm Corning crystallization dish made of PYREX glass. Rims are reinforced and fire polished to reduce chipping. *Photo depicts default sizing. 8465-20 (509558483) is 170 x 90mm
This item is not returnable. View return policy
Catalog No. 50-955-8483 Supplier Ace Glass, Inc. Supplier No. 846520
170X90mm Corning crystallization dish made of PYREX glass. Rims are reinforced and fire polished to reduce chipping. *Photo depicts default sizing. 8465-20 (509558483) is 170 x 90mm